Assay Details
| Gene Symbol: Hectd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175244.3 | NP_780453.1 | 5 - 6 | 930 | 96 | |
| GenBank mRNA | AK051385.1 | - | 5 - 6 | 943 | 96 | |
| AK139305.1 | - | 5 - 6 | 958 | 96 | ||
| AK154378.1 | - | 5 - 6 | 962 | 96 | ||
| AK155290.1 | - | 5 - 6 | 943 | 96 | ||
| AK155676.1 | - | 5 - 6 | 943 | 96 | ||
| AK163323.1 | - | 5 - 6 | 965 | 96 | ||
| AK165482.1 | - | 5 - 6 | 962 | 96 | ||
| AK170930.1 | - | 5 - 6 | 943 | 96 | ||
| BC070411.1 | - | 5 - 6 | 930 | 5706209 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hectd3
Genomic Map