Assay Details
| Gene Symbol: Nhlrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175340.4 | NP_780549.1 | 1 | 2187 | 61 | |
| GenBank mRNA | AK045746.1 | - | 1 | 2158 | 61 | |
| BC062644.1 | - | 1 | 2156 | 61 | ||
| BX514773.1 | - | 1 | 330 | 61 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
autophagy
protein ubiquitination
positive regulation of protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
protein binding
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity
protein polyubiquitination
autophagy
protein ubiquitination
positive regulation of protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
protein binding
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29011093 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29011121 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29011113 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29011115 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29011114 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29011119 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29011120 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29011116 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P20727 | BX514773.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P20727 | BC062644.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P20727 | NM_175340.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P20727 | AK045746.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P20727 | BX514773.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P20727 | BC062644.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P20727 | NM_175340.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P20727 | AK045746.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044847 | BC062644.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044847 | BC118951.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044847 | BC118952.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044847 | NM_175340.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044847 | AK045746.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 466374 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129434 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169910 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133065 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044847 | BC062644.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044847 | BC118951.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044847 | BC118952.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044847 | NM_175340.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044847 | AK045746.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P20727 | BX514773.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P20727 | BC062644.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P20727 | NM_175340.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P20727 | AK045746.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P20727 | BX514773.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P20727 | BC062644.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P20727 | NM_175340.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P20727 | AK045746.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P20727 | BX514773.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P20727 | BC062644.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P20727 | NM_175340.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P20727 | AK045746.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P20727 | BX514773.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P20727 | BC062644.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P20727 | NM_175340.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P20727 | AK045746.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29011108 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169910 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129434 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133065 | - | - |
| Whole Mouse Genome G4122A | A_52_P20727 | BX514773.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P20727 | BC062644.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P20727 | NM_175340.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P20727 | AK045746.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P20727 | BX514773.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P20727 | BC062644.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P20727 | NM_175340.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P20727 | AK045746.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104267 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129434 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169910 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133065 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10834264 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10680233 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11017943 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10680234 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29011093 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29011115 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29011116 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29011119 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29011120 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29011114 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29011121 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29011113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29011053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10753312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10753313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011140 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10575100 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011113 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10651194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10703830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10842623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11017943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011143 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11017932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10992503 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10687701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10865603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011097 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011084 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11094278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29011155 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10409019 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409017 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409018 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409020 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435213_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10409016 | - | - |
| Mouse Genome 430 2.0 Array | 1435213_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435213_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 105193_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 105193_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300011964.mm.1 | NM_175340.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300011964.mm.1 | NM_175340.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300011964.mm.1 | TC1300001926.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435213_PM_at | - | - |
| Mouse Expression Set 430 | 1435213_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435213_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435213_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 663590 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1450128 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001450128 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2595091 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2595091 | - | - |
| mouse-6 v1.1 (Illumina) | scl44021.1.1_258-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1450128 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl44021.1.1_258-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1450128 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2595091 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2595091 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl44021.1.1_258-S | - | - |

Nhlrc1
Genomic Map