| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100009903.mm.1 |
NM_175443.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009903.mm.1 |
NM_175443.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009903.mm.1 |
TC0100001207.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100009907.mm.1 |
NM_175443.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009907.mm.1 |
NM_175443.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009907.mm.1 |
TC0100001207.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100009906.mm.1 |
NM_175443.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009906.mm.1 |
NM_175443.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009906.mm.1 |
TC0100001207.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100009902.mm.1 |
NM_175443.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009902.mm.1 |
NM_175443.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009902.mm.1 |
TC0100001207.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100009904.mm.1 |
NM_175443.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009904.mm.1 |
NM_175443.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009904.mm.1 |
TC0100001207.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100009909.mm.1 |
NM_175443.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009909.mm.1 |
NM_175443.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009909.mm.1 |
TC0100001207.mm.1 |
MTA TC |