Assay Details
| Gene Symbol: Nyap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB429289.1 | - | 3 - 4 | 2215 | 69 | |
| AK032567.1 | - | 1 - 2 | 1388 | 69 | ||
| AK080821.1 | - | 4 - 5 | 2585 | 69 | ||
| AK141276.1 | - | 4 - 5 | 2072 | 69 | ||
| AK148980.1 | - | 1 - 2 | 1179 | 69 | ||
More Information
Set Membership: |
|

Nyap1
Genomic Map