Assay Details
| Gene Symbol: Nyap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175521.3 | NP_780730.2 | 5 - 6 | 2526 | 137 | |
| GenBank mRNA | AB429289.1 | - | 5 - 6 | 2523 | 137 | |
| AK032567.1 | - | 3 - 4 | 1696 | 137 | ||
| AK080821.1 | - | 6 - 7 | 2893 | 137 | ||
| AK148980.1 | - | 3 - 4 | 1487 | 137 | ||
| BC057372.1 | - | 5 - 6 | 2526 | 5686602 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24127317 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24127314 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24127315 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24127313 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24127312 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24127316 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999923 | BC057372.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999923 | AK148980.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999923 | AK032567.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999923 | AK080821.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999923 | NM_175521.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348182 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 659311 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 30168 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 29721 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 448320 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 661470 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559615 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 150325 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152716 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132204 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999923 | BC057372.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999923 | AK148980.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999923 | AK032567.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999923 | AK080821.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999923 | NM_175521.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127306 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24127320 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152716 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132204 | - | - |
| Whole Mouse Genome G4122A | A_52_P279579 | BC057372.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P279579 | AK148980.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P279579 | AK032567.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P279579 | AK080821.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P279579 | NM_175521.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110604 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152716 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132204 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04422065 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04422068 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04321152 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04321153 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04422069 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04321154 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04885661 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04422074 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04422067 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04102519 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24127312 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24127313 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24127314 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24127317 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24127316 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24127315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24127265 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127317 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04422065 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04604738 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04422074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04645482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127320 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04146057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127303 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04645498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127314 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127257 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04688023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127261 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04645492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04519459 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127265 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04615314 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04645489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04897897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04102520 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127278 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127259 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24127306 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04422067 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10534915 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534918 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534913 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534917 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534919 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534914 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534916 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439770_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10534912 | - | - |
| Mouse Genome 430 2.0 Array | 1439770_at | - | - |
| Mouse Genome 430 2.0 Array | 1439770_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 243300_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 243300_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500031534.mm.1 | NM_175521.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031534.mm.1 | NM_175521.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031534.mm.1 | TC0500003461.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439770_at | - | - |
| Mouse Expression Set 430 | 1439770_PM_at | - | - |
| Murine Genome U74v2 Set | 141048_PM_at | - | - |
| Murine Genome U74v2 Set | 141048_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439770_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439770_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 589069 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6130546 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006130546 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2621130 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2621130 | - | - |
| mouse-6 v1.1 (Illumina) | scl0243300.1_92-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6130546 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0243300.1_92-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6130546 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2621130 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2621130 | - | - |

Nyap1
Genomic Map