Assay Details
| Gene Symbol: Gldn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177350.5 | NP_796324.1 | 3 - 4 | 470 | 75 | |
| GenBank mRNA | AF548022.1 | - | 3 - 4 | 126 | 75 | |
| AK031523.1 | - | 3 - 4 | 471 | 75 | ||
| BC125025.1 | - | 3 - 4 | 450 | 40058551 | 75 | |
| BC125026.1 | - | 3 - 4 | 450 | 40058552 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gldn
Genomic Map