Assay Details
| Gene Symbol: Gldn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177350.5 | NP_796324.1 | 7 - 8 | 926 | 74 | |
| GenBank mRNA | AF548022.1 | - | 7 - 8 | 582 | 74 | |
| AK031523.1 | - | 7 - 8 | 927 | 74 | ||
| BC125025.1 | - | 7 - 8 | 906 | 40058551 | 74 | |
| BC125026.1 | - | 7 - 8 | 906 | 40058552 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gldn
Genomic Map