Assay Details
| Gene Symbol: Sh3pxd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177364.3 | NP_796338.2 | 2 - 3 | 245 | 74 | |
| GenBank mRNA | AB430861.1 | - | 2 - 3 | 167 | 74 | |
| AK042261.1 | - | 2 - 3 | 231 | 74 | ||
| AK043324.1 | - | 2 - 3 | 217 | 74 | ||
| AK089330.1 | - | 2 - 3 | 217 | 74 | ||
| AK090119.1 | - | 2 - 3 | 238 | 74 | ||
| AK154898.1 | - | 2 - 3 | 203 | 74 | ||
| AK163471.1 | - | 2 - 3 | 217 | 74 | ||
| AK169978.1 | - | 2 - 3 | 237 | 74 | ||
| AK171384.1 | - | 2 - 3 | 217 | 74 | ||
| AK220175.1 | - | 2 - 3 | 234 | 74 | ||
| BC115711.1 | - | 2 - 3 | 214 | 74 | ||
| BC115764.1 | - | 2 - 3 | 214 | 74 | ||
| BC141306.1 | - | 2 - 3 | 217 | 9055918 | 74 | |
| BY197208.1 | - | 2 - 3 | 245 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3pxd2b
Genomic Map