Assay Details
| Gene Symbol: Mia3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177389.3 | NP_796363.2 | 1 - 2 | 164 | 87 | |
| GenBank mRNA | AK044749.1 | - | 1 - 2 | 141 | 87 | |
| AK084344.1 | - | 1 - 2 | 153 | 87 | ||
| AK148470.1 | - | 1 - 2 | 153 | 87 | ||
| BB618500.1 | - | 1 - 2 | 164 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
chondrocyte development
positive regulation of leukocyte migration
transport
exocytosis
negative regulation of cell adhesion
protein transport
vesicle-mediated transport
collagen fibril organization
negative regulation of cell migration
positive regulation of bone mineralization
wound healing
molecular_function
chondrocyte development
positive regulation of leukocyte migration
transport
exocytosis
negative regulation of cell adhesion
protein transport
vesicle-mediated transport
collagen fibril organization
negative regulation of cell migration
positive regulation of bone mineralization
wound healing
molecular_function
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139460 | BM239089.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139460 | NM_177389.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P139460 | AK046506.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P315085 | AK220252.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P315085 | BC012393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P315085 | BC012632.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P315085 | BC038889.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P315085 | BC125472.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P315085 | AK220252.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P315085 | BC012393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P315085 | BC012632.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P315085 | BC038889.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P315085 | BC125472.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P315085 | AK220252.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P315085 | BC012393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P315085 | BC012632.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P315085 | BC038889.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P315085 | BC125472.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P315085 | AK220252.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P315085 | BC012393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P315085 | BC012632.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P315085 | BC038889.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P315085 | BC125472.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139460 | BM239089.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139460 | NM_177389.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P139460 | AK046506.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09574558 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09574537 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09574541 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09574547 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09574566 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09574570 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09574552 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09574561 | - | - |
| Whole Mouse Genome G4122A | A_52_P637034 | AK220252.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P637034 | BC125472.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P637034 | NM_177389.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P315085 | AK220252.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P315085 | BC012393.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P315085 | BC012632.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P315085 | BC038889.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P315085 | BC125472.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P315085 | AK220252.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P315085 | BC012393.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P315085 | BC012632.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P315085 | BC038889.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P315085 | BC125472.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P259658 | NM_177389.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P259658 | AK044749.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136592 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09574561 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09574537 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10598200 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598201 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598199 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598202 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1459984_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434698_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455066_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10598198 | - | - |
| Mouse Genome 430 2.0 Array | 1459070_at | - | - |
| Mouse Genome 430 2.0 Array | 1459984_at | - | - |
| Mouse Genome 430 2.0 Array | 1459070_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1459984_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100030560.mm.1 | NM_177389.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100030560.mm.1 | NM_177389.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100030560.mm.1 | TC0100003725.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100030562.mm.1 | NM_177389.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100030562.mm.1 | NM_177389.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100030562.mm.1 | TC0100003725.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1459070_PM_at | - | - |
| Mouse Expression Set 430 | 1459070_at | - | - |
| Mouse Expression Set 430 | 1459984_at | - | - |
| Mouse Expression Set 430 | 1459984_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455066_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434698_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455066_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434698_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459984_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459984_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 405010 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5550746 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5130711 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3710092 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005130711 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005550746 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003710092 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2692545 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2689331 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2652104 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1226269 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2600646 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1258474 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2692545 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226269 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2600646 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2689331 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2652104 | - | - |
| mouse-6 v1.1 (Illumina) | scl0338366.1_12-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00338366.1_62-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00338366.1_25-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5130711 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5550746 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3710092 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00338366.1_62-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0338366.1_12-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00338366.1_25-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5550746 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5130711 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3710092 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2600646 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1226269 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2600646 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226269 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2689331 | - | - |

Mia3
Genomic Map