Assay Details
| Gene Symbol: H2-M11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177635.1 | NP_808303.1 | 1 - 2 | 78 | 70 | |
| GenBank mRNA | AF539596.1 | - | 1 - 2 | 78 | 70 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

H2-M11
Genomic Map