Assay Details
| Gene Symbol: Gskip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178613.3 | NP_848728.2 | 2 - 3 | 364 | 57 | |
| GenBank mRNA | AK029484.1 | - | 2 - 3 | 276 | 57 | |
| AK042664.1 | - | 2 - 3 | 325 | 57 | ||
| AK079775.1 | - | 2 - 3 | 310 | 57 | ||
| AK168962.1 | - | 2 - 3 | 328 | 57 | ||
| AK171097.1 | - | 2 - 3 | 341 | 57 | ||
| BC031494.1 | - | 2 - 3 | 332 | 3156862 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28710690 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28710688 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28710687 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28710689 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2000329 | AK029484.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2000329 | AK030204.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2000329 | NM_178613.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 124180 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 839830 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 944480 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576753 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 493202 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 749787 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 958462 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 21803 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 774928 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 134016 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 161953 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 958176 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52756 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 104425 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109938 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141818 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2000329 | AK029484.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2000329 | AK030204.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2000329 | NM_178613.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28710700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28710702 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28710697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28710696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28710705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28710689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28710701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28710695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28710698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28710704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28710691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28710694 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02597456 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07737777 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141818 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109938 | - | - |
| Whole Mouse Genome G4122A | A_52_P241987 | AK079775.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P241987 | AK168962.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P241987 | AK171097.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P241987 | AK029484.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P241987 | AK030204.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P332917 | AK029484.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P332917 | AK030204.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P332917 | NM_178613.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P338039 | BC031494.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P338039 | AK079775.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P338039 | AK168962.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P338039 | AK171097.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P338039 | AK029484.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108195 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141818 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109938 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10416686 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10416691 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28710688 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28710687 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28710690 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28710689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28710735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10330293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09934707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10000238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710802 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09991482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10384226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10171443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710754 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10271983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710736 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710770 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710800 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710731 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10358967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710786 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710811 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710784 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10416686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710790 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710726 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710807 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710774 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09991487 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710727 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710714 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10384230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710733 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09991488 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10107518 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09822681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710702 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09916622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10384223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10359705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10416691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28710695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10404740 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10398125 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398127 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398129 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398126 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398128 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431334_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435423_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435422_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433700_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10398124 | - | - |
| Mouse Genome 430 2.0 Array | 1435422_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431334_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1435423_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1433700_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1433700_at | - | - |
| Mouse Genome 430 2.0 Array | 1435423_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1435422_at | - | - |
| Mouse Genome 430 2.0 Array | 1431334_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66787_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66787_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200007625.mm.1 | NM_178613.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007625.mm.1 | NM_178613.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007625.mm.1 | TC1200001070.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007626.mm.1 | NM_178613.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007626.mm.1 | NM_178613.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007626.mm.1 | TC1200001070.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007627.mm.1 | NM_178613.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007627.mm.1 | NM_178613.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007627.mm.1 | TC1200001070.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1433700_at | - | - |
| Mouse Expression Set 430 | 1435423_x_at | - | - |
| Mouse Expression Set 430 | 1433700_PM_at | - | - |
| Mouse Expression Set 430 | 1431334_a_at | - | - |
| Mouse Expression Set 430 | 1435422_at | - | - |
| Mouse Expression Set 430 | 1435422_PM_at | - | - |
| Mouse Expression Set 430 | 1431334_PM_a_at | - | - |
| Mouse Expression Set 430 | 1435423_PM_x_at | - | - |
| Murine Genome U74v2 Set | 132751_i_at | - | - |
| Murine Genome U74v2 Set | 132751_PM_i_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435422_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435423_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433700_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431334_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431334_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435422_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435423_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433700_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 861059 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 104200292 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 510592 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104200609 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100460377 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000510592 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237665 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1250795 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237665 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1260181 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1229851 | - | - |
| mouse-6 v1.1 (Illumina) | scl42813.4_28-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 510592 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104200609 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100460377 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104200292 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl42813.4_28-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104200609 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100460377 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 510592 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104200292 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1237665 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2551506 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1250795 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237665 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1260181 | - | - |

Gskip
Genomic Map