| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200028881.mm.1 |
NM_001243102.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028881.mm.1 |
NM_001243102.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028881.mm.1 |
NM_001243103.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028881.mm.1 |
NM_001243103.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028881.mm.1 |
NM_178885.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028881.mm.1 |
NM_178885.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028881.mm.1 |
TC0200003359.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200028882.mm.1 |
NM_001243102.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028882.mm.1 |
NM_001243102.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028882.mm.1 |
NM_001243103.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028882.mm.1 |
NM_001243103.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028882.mm.1 |
NM_178885.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028882.mm.1 |
NM_178885.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028882.mm.1 |
TC0200003359.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200028883.mm.1 |
NM_001243102.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028883.mm.1 |
NM_001243102.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028883.mm.1 |
NM_001243103.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028883.mm.1 |
NM_001243103.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028883.mm.1 |
NM_178885.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028883.mm.1 |
NM_178885.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028883.mm.1 |
TC0200003359.mm.1 |
MTA TC |