Assay Details
| Gene Symbol: Ushbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181418.3 | NP_852083.2 | 1 - 2 | 93 | 65 | |
| GenBank mRNA | AK034939.1 | - | 1 - 2 | 95 | 65 | |
| AK043798.1 | - | 1 - 2 | 83 | 65 | ||
| BC026453.1 | - | 1 - 2 | 77 | 4460920 | 65 | |
| BY241807.1 | - | 1 - 2 | 109 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC026453.1, NM_181418.3 transcripts.
Set Membership: |
|

Ushbp1
Genomic Map