Assay Details
| Gene Symbol: Ushbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181418.3 | NP_852083.2 | 4 - 5 | 779 | 66 | |
| GenBank mRNA | AK034939.1 | - | 3 - 4 | 949 | 66 | |
| AK043798.1 | - | 4 - 5 | 769 | 66 | ||
| AK084049.1 | - | 1 - 2 | 226 | 66 | ||
| BC026453.1 | - | 4 - 5 | 763 | 4460920 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ushbp1
Genomic Map