Assay Details
| Gene Symbol: Ushbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181418.3 | NP_852083.2 | 6 - 7 | 76 | ||
| GenBank mRNA | AK034939.1 | - | 5 - 6 | 76 | ||
| AK084049.1 | - | 3 - 4 | 76 | |||
| BC026453.1 | - | 6 - 7 | 1021 | 4460920 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ushbp1
Genomic Map