Assay Details
| Gene Symbol: Pkd2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181422.3 | NP_852087.2 | 5 - 6 | 1126 | 68 | |
| GenBank mRNA | AB290927.1 | - | 5 - 6 | 958 | 68 | |
| AF271381.1 | - | 5 - 6 | 1092 | 68 | ||
| AK046772.1 | - | 5 - 6 | 1127 | 68 | ||
| BC116297.2 | - | 5 - 6 | 1057 | 40094997 | 68 | |
| BC116323.2 | - | 5 - 6 | 1057 | 40096335 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pkd2l1
Genomic Map