Assay Details
| Gene Symbol: Pkd2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181422.3 | NP_852087.2 | 6 - 7 | 1349 | 81 | |
| GenBank mRNA | AB290927.1 | - | 6 - 7 | 1181 | 81 | |
| AF271381.1 | - | 6 - 7 | 1315 | 81 | ||
| AK046772.1 | - | 6 - 7 | 1350 | 81 | ||
| BC116297.2 | - | 6 - 7 | 1280 | 40094997 | 81 | |
| BC116323.2 | - | 6 - 7 | 1280 | 40096335 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pkd2l1
Genomic Map