Assay Details
| Gene Symbol: Dennd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146122.3 | NP_666234.3 | 12 - 13 | 1116 | 78 | |
| GenBank mRNA | AK136192.1 | - | 5 - 6 | 401 | 78 | |
| AK137530.1 | - | 11 - 12 | 1044 | 78 | ||
| AK220407.1 | - | 10 - 11 | 864 | 78 | ||
| BC027786.1 | - | 12 - 13 | 1079 | 5346095 | 78 | |
| DQ448594.1 | - | 12 - 13 | 861 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dennd1a
Genomic Map