Assay Details
| Gene Symbol: Tmem44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172614.3 | NP_766202.3 | 1 - 2 | 219 | 66 | |
| GenBank mRNA | AK031524.1 | - | 1 - 2 | 181 | 66 | |
| AK048655.1 | - | 1 - 2 | 220 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK048655.1 transcripts.
Set Membership: |
|

Tmem44
Genomic Map