Assay Details
| Gene Symbol: Insc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173767.3 | NP_776128.2 | 6 - 7 | 980 | 102 | |
| GenBank mRNA | AK014452.1 | - | 5 - 6 | 741 | 102 | |
| AK050143.1 | - | 3 - 4 | 475 | 102 | ||
| BC026405.1 | - | 6 - 7 | 867 | 4211657 | 102 | |
| BC089586.1 | - | 3 - 4 | 484 | 102 | ||
| BC139196.1 | - | 3 - 4 | 368 | 8862218 | 102 | |
| BC139197.1 | - | 3 - 4 | 368 | 8862219 | 102 | |
| DQ205645.1 | - | 5 - 6 | 693 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Insc
Genomic Map