Assay Details
| Gene Symbol: Insc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173767.3 | NP_776128.2 | 10 - 11 | 1523 | 60 | |
| GenBank mRNA | AK050143.1 | - | 7 - 8 | 1018 | 60 | |
| AK178410.1 | - | 2 - 3 | 197 | 60 | ||
| BC026405.1 | - | 10 - 11 | 1410 | 4211657 | 60 | |
| BC089586.1 | - | 7 - 8 | 1027 | 60 | ||
| BC139196.1 | - | 7 - 8 | 911 | 8862218 | 60 | |
| BC139197.1 | - | 7 - 8 | 911 | 8862219 | 60 | |
| DQ205645.1 | - | 9 - 10 | 1236 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Insc
Genomic Map