Assay Details
Gene Symbol: Lrrc28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027413.1 | NP_081689.1 | 1 - 2 | 56 | 78 | |
NM_175124.5 | NP_780333.3 | 1 - 2 | 56 | 78 | ||
NR_028143.1 | 1 - 2 | 56 | 78 | |||
NR_028144.1 | 1 - 2 | 56 | 78 | |||
GenBank mRNA | AI663251.1 | - | 1 - 2 | 56 | 78 | |
AK077270.1 | - | 1 - 2 | 52 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK077270.1, NM_027413.1, NM_175124.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
