Assay Details
| Gene Symbol: Lrrc28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027413.1 | NP_081689.1 | 4 - 5 | 381 | 102 | |
| NM_175124.5 | NP_780333.3 | 4 - 5 | 381 | 102 | ||
| NR_028143.1 | 4 - 5 | 381 | 102 | |||
| GenBank mRNA | AI663251.1 | - | 4 - 5 | 381 | 102 | |
| AK004904.1 | - | 4 - 5 | 350 | 102 | ||
| AK008713.1 | - | 4 - 5 | 349 | 102 | ||
| AK050154.1 | - | 4 - 5 | 353 | 102 | ||
| AK077270.1 | - | 4 - 5 | 377 | 102 | ||
| AK159417.1 | - | 4 - 5 | 350 | 102 | ||
| BC086694.1 | - | 4 - 5 | 272 | 30697866 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc28
Genomic Map