Assay Details
| Gene Symbol: Ccdc158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177230.3 | NP_796204.1 | 9 - 10 | 1216 | 70 | |
| GenBank mRNA | AK029997.1 | - | 9 - 10 | 1216 | 70 | |
| BC137976.1 | - | 9 - 10 | 1185 | 8860995 | 70 | |
| BC145072.1 | - | 9 - 10 | 1186 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc158
Genomic Map