Assay Details
| Gene Symbol: Negr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039094.3 | NP_001034183.1 | 4 - 5 | 1056 | 73 | |
| NM_177274.4 | NP_796248.1 | 4 - 5 | 1056 | 73 | ||
| GenBank mRNA | AJ487032.1 | - | 4 - 5 | 676 | 73 | |
| AK030503.1 | - | 4 - 5 | 882 | 73 | ||
| AK035218.1 | - | 4 - 5 | 892 | 73 | ||
| AK122576.2 | - | 3 - 4 | 399 | 73 | ||
| AK142298.1 | - | 4 - 5 | 750 | 73 | ||
| AK147253.1 | - | 4 - 5 | 1057 | 73 | ||
| BC146005.1 | - | 4 - 5 | 841 | 40131308 | 73 | |
| BC146007.1 | - | 4 - 5 | 840 | 40131310 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Negr1
Genomic Map