Assay Details
| Gene Symbol: Slx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177472.5 | NP_803423.2 | 12 - 13 | 4364 | 74 | |
| GenBank mRNA | AK041127.1 | - | 1 - 2 | 615 | 74 | |
| AK042952.1 | - | 1 - 2 | 130 | 74 | ||
| AK173263.1 | - | 11 - 12 | 3777 | 74 | ||
| BC042798.1 | - | 2 - 3 | 627 | 74 | ||
| BC065125.1 | - | 12 - 13 | 4369 | 30534878 | 74 | |
| BC082337.1 | - | 5 - 6 | 2855 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slx4
Genomic Map