Assay Details
| Gene Symbol: BC049762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177567.3 | NP_808235.1 | 2 - 3 | 480 | 87 | |
| GenBank mRNA | AK076829.1 | - | 2 - 3 | 481 | 87 | |
| AK154967.1 | - | 3 - 4 | 664 | 87 | ||
| BC049762.1 | - | 2 - 3 | 357 | 6774041 | 87 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27792928 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27792929 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052106 | BC049762.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052106 | NM_177567.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052106 | AK076829.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052106 | AK154967.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093844 | NM_177567.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093844 | AK076829.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093844 | AK154967.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 120582 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524486 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 671215 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 69614 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 876969 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 136768 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128178 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052106 | BC049762.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052106 | NM_177567.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052106 | AK076829.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052106 | AK154967.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093844 | NM_177567.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093844 | AK076829.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093844 | AK154967.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27792904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27792912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27792915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27792905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27792903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27792928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27792922 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27792929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27792908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27792919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27792924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27792910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27792920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27792911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27792916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27792926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27792907 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128178 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07339946 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07339957 | - | - |
| Whole Mouse Genome G4122A | A_52_P573517 | BC049762.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P573517 | NM_177567.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P573517 | AK076829.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P573517 | AK154967.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129308 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128178 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09718568 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09068206 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09718570 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09718580 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27792928 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27792929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27792879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792887 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09203587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09587500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792897 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08975693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08975698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792899 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792893 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27792895 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10385682 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385683 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385685 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385684 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436433_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10385681 | - | - |
| Mouse Genome 430 2.0 Array | 1436433_at | - | - |
| Mouse Genome 430 2.0 Array | 1436433_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 193286_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 193286_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100024563.mm.1 | TC1100002690.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100024559.mm.1 | NM_177567.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024559.mm.1 | NM_177567.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024559.mm.1 | TC1100002690.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436433_at | - | - |
| Mouse Expression Set 430 | 1436433_PM_at | - | - |
| Murine Genome U74v2 Set | 168626_r_at | - | - |
| Murine Genome U74v2 Set | 168626_PM_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436433_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436433_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 323786 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2230563 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2230563 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002230563 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225817 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225817 | - | - |
| mouse-6 v1.1 (Illumina) | scl40261.5.1_18-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2230563 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40261.5.1_18-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2230563 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225817 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225817 | - | - |

BC049762
Genomic Map