Assay Details
| Gene Symbol: Tgm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289747.1 | NP_001276676.1 | 4 - 5 | 653 | 75 | |
| NM_001289748.1 | NP_001276677.1 | 3 - 4 | 490 | 75 | ||
| NM_177726.4 | NP_808394.1 | 3 - 4 | 509 | 75 | ||
| GenBank mRNA | AK037229.1 | - | 3 - 4 | 499 | 75 | |
| AY159126.1 | - | 3 - 4 | 499 | 75 | ||
| AY177607.1 | - | 3 - 4 | 490 | 75 | ||
| BC112420.1 | - | 3 - 4 | 486 | 40061385 | 75 | |
| BC113796.1 | - | 4 - 5 | 630 | 40061392 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC113796.1, NM_001289747.1, NM_001289748.1 transcripts.
Set Membership: |
|

Tgm6
Genomic Map