Assay Details
| Gene Symbol: Ubr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK037622.1 | - | 5 - 6 | 1027 | 80 | |
| BC172146.1 | - | 5 - 6 | 775 | 80 | ||
| DQ924536.1 | - | 5 - 6 | 1086 | 80 | ||
More Information
Set Membership: |
|

Ubr3
Genomic Map