Assay Details
| Gene Symbol: Zfp459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177811.4 | NP_808479.1 | 3 - 4 | 301 | 72 | |
| GenBank mRNA | AK036923.1 | - | 3 - 4 | 302 | 72 | |
| BC100389.1 | - | 3 - 4 | 267 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P452576 | BC100389.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P452576 | NM_177811.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P452576 | AK036923.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P452576 | BC100389.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P452576 | NM_177811.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P452576 | AK036923.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 957120 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206336 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 700340 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 359769 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 809450 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 160515 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11660 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 72254 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172548 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145753 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P452576 | BC100389.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P452576 | NM_177811.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P452576 | AK036923.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P452576 | BC100389.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P452576 | NM_177811.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P452576 | AK036923.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07905666 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145753 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172548 | - | - |
| Whole Mouse Genome G4122A | A_51_P452576 | BC100389.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P452576 | NM_177811.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P452576 | AK036923.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P452576 | BC100389.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P452576 | NM_177811.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P452576 | AK036923.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138889 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172548 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145753 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10886790 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10886792 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10538704 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10538711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29108598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10983818 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108502 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11030749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108503 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108596 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10507773 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10855047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10836803 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10855043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108505 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108631 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108629 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11025480 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10538717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108586 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29108609 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10410318 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410319 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410320 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405917 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10410317 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300014636.mm.1 | NM_177811.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300014636.mm.1 | NM_177811.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300014636.mm.1 | TC1300002247.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300014637.mm.1 | NM_177811.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300014637.mm.1 | NM_177811.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300014637.mm.1 | TC1300002247.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300014635.mm.1 | NM_177811.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300014635.mm.1 | NM_177811.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300014635.mm.1 | TC1300002247.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300014634.mm.1 | NM_177811.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300014634.mm.1 | NM_177811.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300014634.mm.1 | TC1300002247.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300014639.mm.1 | NM_177811.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300014639.mm.1 | NM_177811.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300014639.mm.1 | TC1300002247.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 458763 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 7040215 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 7040215 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007040215 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1253331 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1253331 | - | - |
| mouse-6 v1.1 (Illumina) | scl43800.4_110-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7040215 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl43800.4_110-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7040215 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1253331 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1253331 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl43800.4_110-S | - | - |

Zfp459
Genomic Map