Assay Details
| Gene Symbol: E130311K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177856.4 | NP_808524.1 | 1 - 2 | 130 | 72 | |
| GenBank mRNA | AK087526.1 | - | 1 - 2 | 131 | 72 | |
| BC125500.1 | - | 1 - 2 | 217 | 40130015 | 72 | |
| BC132160.1 | - | 1 - 2 | 217 | 40130437 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

E130311K13Rik
Genomic Map