Assay Details
| Gene Symbol: Vps37b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177876.4 | NP_808544.1 | 1 - 2 | 161 | 70 | |
| GenBank mRNA | AK082658.1 | - | 1 - 2 | 122 | 70 | |
| AK149893.1 | - | 1 - 2 | 128 | 70 | ||
| BC026744.1 | - | 1 - 2 | 162 | 4163734 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vps37b
Genomic Map