Assay Details
| Gene Symbol: Zfp786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177882.4 | NP_808550.1 | 3 - 4 | 400 | 79 | |
| GenBank mRNA | AK080438.1 | - | 3 - 4 | 400 | 79 | |
| BC138897.1 | - | 3 - 4 | 341 | 8861919 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24424520 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24424495 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24424497 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24424496 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24424498 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24424494 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171802 | NM_177882.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171802 | AK080438.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913354 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 804030 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 661355 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 797345 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 458730 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 445775 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 118145 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106634 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139699 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171802 | NM_177882.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171802 | AK080438.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P503297 | NM_177882.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P503297 | AK080438.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24424487 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139699 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106634 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05888857 | - | - |
| Whole Mouse Genome G4122A | A_51_P503297 | NM_177882.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P503297 | AK080438.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136809 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106634 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139699 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05618463 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05021955 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05021953 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05818533 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05618465 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05618455 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05266013 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24424494 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24424520 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24424496 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24424495 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24424498 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24424497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24424471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424475 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05049273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05792930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05792927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05747199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05469786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05747198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05792929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05747218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05049272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05184392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05618468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05747202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05362507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24424483 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10544543 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544546 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544545 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544544 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458214_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10544542 | - | - |
| Mouse Genome 430 2.0 Array | 1458214_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1458214_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 330301_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 330301_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600017393.mm.1 | NM_177882.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600017393.mm.1 | NM_177882.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600017393.mm.1 | TC0600002214.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600017394.mm.1 | NM_177882.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600017394.mm.1 | NM_177882.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600017394.mm.1 | TC0600002214.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600017395.mm.1 | NM_177882.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600017395.mm.1 | NM_177882.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600017395.mm.1 | TC0600002214.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1458214_PM_at | - | - |
| Mouse Expression Set 430 | 1458214_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458214_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458214_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 877144 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4540040 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004540040 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1247019 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1247019 | - | - |
| mouse-6 v1.1 (Illumina) | scl29053.4_277-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4540040 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29053.4_277-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4540040 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1247019 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2876265 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1247019 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2876265 | - | - |

Zfp786
Genomic Map