Assay Details
| Gene Symbol: Tmem150b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177887.4 | NP_808555.1 | 4 - 5 | 602 | 70 | |
| GenBank mRNA | AK041846.1 | - | 4 - 5 | 603 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24950089 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24950090 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24950091 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094499 | AK041846.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094499 | NM_177887.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094494 | BC022651.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094494 | NM_001142792.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117041 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 384749 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 109249 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 145177 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 444809 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 60632 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133267 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170123 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118082 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094499 | AK041846.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094499 | NM_177887.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094494 | BC022651.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094494 | NM_001142792.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950160 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170123 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159176 | - | - |
| Whole Mouse Genome G4122A | A_52_P252070 | AK041846.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P252070 | BC022651.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P252070 | NM_001142792.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P252070 | NM_177887.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P304971 | AK041846.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P304971 | NM_177887.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129049 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170123 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159176 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06102617 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24950090 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24950091 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24950089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06266153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06618825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06102597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06102603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06376818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06102602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06376819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06249226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06102598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06102617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06447344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06249227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06058730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06102607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06102605 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10559647 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559645 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559646 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549694 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549696 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559648 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549695 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549693 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451836_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10549679 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559644 | - | - |
| Mouse Genome 430 2.0 Array | 1451836_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451836_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700019799.mm.1 | NM_001142792.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019799.mm.1 | NM_001142792.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019799.mm.1 | NM_177887.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019799.mm.1 | NM_177887.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019799.mm.1 | TC0700002168.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019800.mm.1 | NM_177887.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019800.mm.1 | NM_177887.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019800.mm.1 | TC0700002168.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019796.mm.1 | NM_001142792.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019796.mm.1 | NM_001142792.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019796.mm.1 | NM_177887.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019796.mm.1 | NM_177887.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019796.mm.1 | TC0700002168.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019792.mm.1 | NM_177887.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019792.mm.1 | NM_177887.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019792.mm.1 | TC0700002168.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019798.mm.1 | NM_001142792.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019798.mm.1 | NM_001142792.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019798.mm.1 | NM_177887.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019798.mm.1 | NM_177887.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019798.mm.1 | TC0700002168.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019797.mm.1 | NM_001142792.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019797.mm.1 | NM_001142792.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019797.mm.1 | NM_177887.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019797.mm.1 | NM_177887.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019797.mm.1 | TC0700002168.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451836_at | - | - |
| Mouse Expression Set 430 | 1451836_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451836_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451836_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451836_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451836_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 603772 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4200685 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004200685 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1216491 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1216491 | - | - |
| mouse-6 v1.1 (Illumina) | scl31805.6.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4200685 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31805.6.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4200685 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2862148 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2862148 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1216491 | - | - |

Tmem150b
Genomic Map