Assay Details
| Gene Symbol: Teddm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178244.3 | NP_839975.2 | 1 | 294 | 144 | |
| GenBank mRNA | AJ554212.1 | - | 1 | 148 | 144 | |
| BC109332.2 | - | 1 | 147 | 144 | ||
| BC141344.1 | - | 1 | 197 | 9055956 | 144 | |
| BC141345.1 | - | 1 | 198 | 9055957 | 144 | |
| CJ048231.1 | - | 1 | 144 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1970335 | AJ554212.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 146622 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593569 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151216 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1970335 | AJ554212.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729197 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151216 | - | - |
| Whole Mouse Genome G4122A | A_52_P616337 | BC141344.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P616337 | BC141345.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P616337 | CJ048231.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P616337 | BC109332.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P616337 | NM_178244.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133806 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151216 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00347187 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00975739 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00975740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00363452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00975729 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00616870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00720899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00087086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00347184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01010632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729207 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729219 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00347188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729205 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00975735 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00975724 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729192 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10350752 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10350751 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100011468.mm.1 | NM_178244.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011468.mm.1 | NM_178244.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011468.mm.1 | TC0100001386.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100011470.mm.1 | NM_178244.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011470.mm.1 | NM_178244.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011470.mm.1 | TC0100001386.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100011471.mm.1 | NM_178244.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011471.mm.1 | NM_178244.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011471.mm.1 | TC0100001386.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 326140 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1240097 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1240097 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001240097 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1235654 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1235654 | - | - |
| mouse-6 v1.1 (Illumina) | scl17356.1.1_256-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1240097 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl17356.1.1_256-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1240097 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1235654 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1235654 | - | - |

Teddm1a
Genomic Map