Assay Details
| Gene Symbol: Helq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081107.1 | NP_001074576.1 | 5 - 6 | 1473 | 60 | |
| GenBank mRNA | AK050769.1 | - | 4 - 5 | 1271 | 60 | |
| AK052427.1 | - | 5 - 6 | 1433 | 60 | ||
| BC082601.1 | - | 5 - 6 | 1437 | 30662103 | 60 | |
| BC109169.2 | - | 5 - 6 | 1341 | 60 | ||
| BC109170.2 | - | 5 - 6 | 1346 | 40048258 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Helq
Genomic Map