Assay Details
| Gene Symbol: Sgpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009163.3 | NP_033189.2 | 10 - 11 | 1106 | 71 | |
| GenBank mRNA | AK036747.1 | - | 10 - 11 | 994 | 71 | |
| AK037789.1 | - | 10 - 11 | 991 | 71 | ||
| AK043024.1 | - | 10 - 11 | 993 | 71 | ||
| AK049342.1 | - | 10 - 11 | 1014 | 71 | ||
| AK075851.1 | - | 1 - 2 | 91 | 71 | ||
| AK150053.1 | - | 10 - 11 | 1001 | 71 | ||
| AK173137.1 | - | 10 - 11 | 1035 | 71 | ||
| BC026135.1 | - | 10 - 11 | 1052 | 4212293 | 71 | |
| CT010214.1 | - | 9 - 10 | 916 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sgpl1
Genomic Map