Assay Details
| Gene Symbol: Azin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301688.1 | NP_001288617.1 | 2 - 3 | 497 | 86 | |
| NM_018745.5 | NP_061215.1 | 2 - 3 | 497 | 86 | ||
| NR_125913.1 | 2 - 3 | 497 | 86 | |||
| GenBank mRNA | AK049680.1 | - | 2 - 3 | 498 | 86 | |
| AK088112.1 | - | 2 - 3 | 495 | 86 | ||
| AK133914.1 | - | 2 - 3 | 494 | 86 | ||
| BC019412.1 | - | 2 - 3 | 485 | 5151406 | 86 | |
| BC021152.1 | - | 2 - 3 | 449 | 86 | ||
| BC046814.1 | - | 2 - 3 | 400 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK049680.1, AK088112.1, BC019412.1, NM_001301688.1, NM_018745.5 transcripts.
Set Membership: |
|

Azin1
Genomic Map