Assay Details
| Gene Symbol: Azin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102458.1 | NP_001095928.1 | 7 - 8 | 1336 | 142 | |
| NM_001301688.1 | NP_001288617.1 | 6 - 7 | 1176 | 142 | ||
| NM_018745.5 | NP_061215.1 | 6 - 7 | 1176 | 142 | ||
| NR_125913.1 | 6 - 7 | 1328 | 142 | |||
| GenBank mRNA | AF032128.1 | - | 4 - 5 | 602 | 142 | |
| AK049680.1 | - | 6 - 7 | 1177 | 142 | ||
| AK088112.1 | - | 6 - 7 | 1174 | 142 | ||
| AK088671.1 | - | 7 - 8 | 1333 | 142 | ||
| AK133914.1 | - | 6 - 7 | 1172 | 142 | ||
| BC019412.1 | - | 6 - 7 | 1164 | 5151406 | 142 | |
| BC021152.1 | - | 6 - 7 | 1129 | 142 | ||
| BC043722.1 | - | 4 - 5 | 682 | 5321733 | 142 | |
| BC046814.1 | - | 6 - 7 | 1231 | 142 | ||
| CF738062.1 | - | 2 - 3 | 167 | 142 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Azin1
Genomic Map