Assay Details
| Gene Symbol: Olfr812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146795.1 | NP_667006.1 | 1 | 225 | 150 | |
| GenBank mRNA | BC132194.1 | - | 1 | 247 | 40130471 | 150 |
| BC132196.1 | - | 1 | 247 | 40130473 | 150 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022048 | NM_146795.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022048 | BC132194.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022048 | BC132196.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022048 | NM_146795.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022048 | BC132194.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022048 | BC132196.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P456699 | NM_146795.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P456699 | BC132194.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P456699 | BC132196.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547472 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27547473 | - | - |
| Whole Mouse Genome G4122A | A_51_P456699 | NM_146795.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P456699 | BC132194.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P456699 | BC132196.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115694 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08373052 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08373095 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08373081 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08373094 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08373073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27547483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27547485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27547455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27547447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27547472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27547484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27547456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27547469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27547475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27547445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27547479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27547491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27547467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27547489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27547473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27547449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27547453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27547488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27547443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08293379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08410999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08373060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27547475 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10373648 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10373647 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000023967.mm.1 | TC1000003193.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000023968.mm.1 | NM_146795.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023968.mm.1 | NM_146795.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023968.mm.1 | TC1000003193.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 758565 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 102190047 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4200121 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004200121 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2680939 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232617 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2680939 | - | - |
| mouse-6 v1.1 (Illumina) | GI_22129258-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0258791.1_328-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102190047 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4200121 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258791.1_328-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4200121 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102190047 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2680939 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1232617 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2985491 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2680939 | - | - |

Olfr812
Genomic Map