Assay Details
| Gene Symbol: Olfr582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147053.1 | NP_667264.1 | 1 | 639 | 148 | |
| GenBank mRNA | BC107338.1 | - | 1 | 639 | 148 | |
| BC107339.1 | - | 1 | 639 | 148 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P130528 | NM_147053.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P130528 | BC107338.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P130528 | BC107339.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133223 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P130528 | NM_147053.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P130528 | BC107338.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P130528 | BC107339.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381780 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133223 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130528 | NM_147053.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130528 | BC107338.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130528 | BC107339.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139721 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123587 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06664638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06075703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06075702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381757 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10555759 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10555763 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10555758 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10555762 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700012712.mm.1 | NM_147053.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700012712.mm.1 | NM_147053.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700012712.mm.1 | TC0700001474.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 667004 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3170390 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003170390 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2696600 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2696600 | - | - |
| mouse-6 v1.1 (Illumina) | scl0259055.1_328-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3170390 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0259055.1_328-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3170390 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2696600 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2696600 | - | - |

Olfr582
Genomic Map