Assay Details
| Gene Symbol: Rdh19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147222.2 | NP_671755.2 | 2 - 3 | 669 | 148 | |
| GenBank mRNA | AY046408.1 | - | 2 - 3 | 665 | 148 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177194 | AY046408.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177194 | NM_147222.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177194 | AY046408.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177194 | NM_147222.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923587 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 311149 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 772205 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 100456 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687392 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 434731 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 536473 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 162627 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 136281 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151959 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148169 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177194 | AY046408.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177194 | NM_147222.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177194 | AY046408.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177194 | NM_147222.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177194 | AY046408.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177194 | NM_147222.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177194 | AY046408.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177194 | NM_147222.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538306 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538310 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148169 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07231501 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07231489 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151959 | - | - |
| Whole Mouse Genome G4122A | A_51_P177194 | AY046408.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177194 | NM_147222.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177194 | AY046408.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177194 | NM_147222.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132647 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117891 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148169 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151959 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08973267 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08973246 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08973254 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08973250 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08973266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08842520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08842508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08956745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08973260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08544320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08842537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08393720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08973250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08956748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08842505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08783653 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08973267 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10367058 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367056 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367057 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10367055 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000011361.mm.1 | NM_147222.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011361.mm.1 | NM_147222.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011361.mm.1 | TC1000001575.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011365.mm.1 | NM_147222.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011365.mm.1 | NM_147222.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011365.mm.1 | TC1000001575.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011362.mm.1 | NM_147222.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011362.mm.1 | NM_147222.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011362.mm.1 | TC1000001575.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011363.mm.1 | NM_147222.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011363.mm.1 | NM_147222.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011363.mm.1 | TC1000001575.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011360.mm.1 | NM_147222.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011360.mm.1 | NM_147222.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011360.mm.1 | TC1000001575.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011364.mm.1 | NM_147222.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011364.mm.1 | NM_147222.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011364.mm.1 | TC1000001575.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 862542 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 840273 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000840273 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240738 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240738 | - | - |
| mouse-6 v1.1 (Illumina) | scl38306.4.1_151-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 840273 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl38306.4.1_151-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 840273 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1240738 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240738 | - | - |

Rdh19
Genomic Map