Assay Details
| Gene Symbol: Zfp111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019940.2 | NP_064324.2 | 3 - 4 | 307 | 149 | |
| GenBank mRNA | AF167318.1 | - | 1 - 2 | 215 | 149 | |
| BB618616.1 | - | 3 - 4 | 308 | 149 | ||
| BC005456.1 | - | 3 - 4 | 243 | 3490047 | 149 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2059159 | BC005456.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2059159 | AF167318.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2059159 | NM_019940.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114611 | AF167318.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114611 | NM_019940.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795304 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 186488 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 469679 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 290188 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 140713 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 938167 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 739842 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 582038 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 78623 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 250119 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350157 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 960953 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 80634 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 108900 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111472 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159711 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2059159 | BC005456.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2059159 | AF167318.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2059159 | NM_019940.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114611 | AF167318.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114611 | NM_019940.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003888 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159711 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111472 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06140344 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01485039 | - | - |
| Whole Mouse Genome G4122A | A_52_P366443 | BC005456.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P366443 | AF167318.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P366443 | NM_019940.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133851 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111472 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159711 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06037520 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06037519 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06396595 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06396599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06332231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06061125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06003992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06004007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003893 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06627986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06396597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06445738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06003997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06037514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06396598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06555323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003897 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05911623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06253748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003878 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10560822 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560821 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560823 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560825 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498796 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560819 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560820 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560824 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421130_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421131_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457185_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10560818 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10498795 | - | - |
| Mouse Genome 430 2.0 Array | 1421130_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421131_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1421130_at | - | - |
| Mouse Genome 430 2.0 Array | 1457185_at | - | - |
| Mouse Genome 430 2.0 Array | 1421131_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1457185_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56707_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56707_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700022370.mm.1 | NM_019940.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022370.mm.1 | NM_019940.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022370.mm.1 | TC0700002591.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022371.mm.1 | NM_019940.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022371.mm.1 | NM_019940.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022371.mm.1 | TC0700002591.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022369.mm.1 | NM_019940.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022369.mm.1 | NM_019940.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022369.mm.1 | TC0700002591.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022366.mm.1 | NM_019940.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022366.mm.1 | NM_019940.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022366.mm.1 | TC0700002591.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022368.mm.1 | NM_019940.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022368.mm.1 | NM_019940.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022368.mm.1 | TC0700002591.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022367.mm.1 | NM_019940.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022367.mm.1 | NM_019940.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022367.mm.1 | TC0700002591.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022372.mm.1 | NM_019940.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022372.mm.1 | NM_019940.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022372.mm.1 | TC0700002591.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421130_at | - | - |
| Mouse Expression Set 430 | 1421131_a_at | - | - |
| Mouse Expression Set 430 | 1421130_PM_at | - | - |
| Mouse Expression Set 430 | 1421131_PM_a_at | - | - |
| Mouse Expression Set 430 | 1457185_at | - | - |
| Mouse Expression Set 430 | 1457185_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421131_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421130_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421131_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421130_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421130_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421131_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421130_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421131_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457185_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457185_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 645689 | - | - |
| ABI Mouse Genome Survey Microarray | 829371 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5860164 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005860164 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2444468 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2444468 | - | - |
| mouse-6 v1.1 (Illumina) | scl31656.4_8-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5860164 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31656.4_8-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5860164 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2836693 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2836698 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2836693 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2836698 | - | - |

Zfp111
Genomic Map