Assay Details
| Gene Symbol: Rbak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045482.2 | NP_001038947.1 | 4 - 5 | 508 | 150 | |
| NM_021326.3 | NP_067301.1 | 4 - 5 | 608 | 150 | ||
| GenBank mRNA | AF226870.1 | - | 4 - 5 | 466 | 150 | |
| AK051009.1 | - | 4 - 5 | 609 | 150 | ||
| BC059913.1 | - | 2 - 3 | 367 | 150 | ||
| CN699599.1 | - | 4 - 5 | 302 | 150 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24155334 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24155332 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24155328 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24155333 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24155330 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24155329 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24155331 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P192307 | BC059913.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P192307 | AK051009.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P192307 | NM_001045482.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P192307 | NM_021326.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P192307 | BC059913.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P192307 | AK051009.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P192307 | NM_001045482.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P192307 | NM_021326.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 582547 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 388368 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 113371 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 192187 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 507674 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151871 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P192307 | BC059913.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P192307 | AK051009.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P192307 | NM_001045482.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P192307 | NM_021326.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P192307 | BC059913.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P192307 | AK051009.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P192307 | NM_001045482.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P192307 | NM_021326.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P192307 | BC059913.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P192307 | AK051009.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P192307 | NM_001045482.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P192307 | NM_021326.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P192307 | BC059913.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P192307 | AK051009.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P192307 | NM_001045482.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P192307 | NM_021326.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P128725 | BC059913.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P128725 | AF226870.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P128725 | AK051009.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P128725 | NM_001045482.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P128725 | NM_021326.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24155330 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151871 | - | - |
| Whole Mouse Genome G4122A | A_51_P128725 | BC059913.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128725 | AF226870.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128725 | AK051009.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128725 | NM_001045482.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128725 | NM_021326.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P192307 | BC059913.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P192307 | AK051009.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P192307 | NM_001045482.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P192307 | NM_021326.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P192307 | BC059913.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P192307 | AK051009.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P192307 | NM_001045482.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P192307 | NM_021326.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114587 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151871 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04763923 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04763921 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04763931 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04763933 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24155331 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24155333 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24155328 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24155332 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24155329 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24155334 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24155330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24155303 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155325 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04696298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155320 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04648124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04339503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155318 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155307 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155315 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04187476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04960633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155312 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04960627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04240563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04339506 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155317 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155330 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04815627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155303 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24155326 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04648127 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10535423 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535420 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535424 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535419 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535425 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535422 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535421 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455323_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10535418 | - | - |
| Mouse Genome 430 2.0 Array | 1455323_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455323_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57782_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57782_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500032509.mm.1 | NM_001045482.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032509.mm.1 | NM_021326.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032509.mm.1 | TC0500003549.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032508.mm.1 | NM_001045482.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032508.mm.1 | NM_021326.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032508.mm.1 | TC0500003549.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032507.mm.1 | NM_001045482.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032507.mm.1 | NM_021326.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032507.mm.1 | TC0500003549.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455323_PM_at | - | - |
| Mouse Expression Set 430 | 1455323_at | - | - |
| Murine Genome U74v2 Set | 136558_at | - | - |
| Murine Genome U74v2 Set | 136558_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455323_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455323_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 793506 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1770008 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1770008 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001770008 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2626773 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2626773 | - | - |
| mouse-6 v1.1 (Illumina) | scl25810.5_670-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1770008 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25810.5_670-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1770008 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2626773 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3054271 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3130983 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3130983 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2626773 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3054271 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl25810.5_670-S | - | - |

Rbak
Genomic Map