Assay Details
| Gene Symbol: Mtdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026002.4 | NP_080278.3 | 10 - 11 | 1826 | 64 | |
| GenBank mRNA | AF501309.1 | - | 10 - 11 | 1764 | 64 | |
| AK029915.1 | - | 10 - 11 | 1827 | 64 | ||
| AK035302.1 | - | 2 - 3 | 220 | 64 | ||
| AK151269.1 | - | 10 - 11 | 1810 | 64 | ||
| AK151810.1 | - | 10 - 11 | 1821 | 64 | ||
| AY553638.1 | - | 10 - 11 | 1509 | 64 | ||
| BC049700.1 | - | 3 - 4 | 448 | 64 | ||
| BC138858.1 | - | 10 - 11 | 1540 | 8861880 | 64 | |
| BC138859.1 | - | 10 - 11 | 1540 | 8861881 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtdh
Genomic Map