Assay Details
| Gene Symbol: Lsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017405.2 | NP_059101.1 | 3 - 4 | 856 | 68 | |
| GenBank mRNA | BC004672.1 | - | 3 - 4 | 602 | 3500090 | 68 |
| U49507.1 | - | 3 - 4 | 747 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lsr
Genomic Map