Assay Details
| Gene Symbol: Lsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164184.1 | NP_001157656.1 | 4 - 5 | 1000 | 60 | |
| NM_017405.2 | NP_059101.1 | 5 - 6 | 1057 | 60 | ||
| GenBank mRNA | AK146807.1 | - | 4 - 5 | 716 | 60 | |
| BC004672.1 | - | 5 - 6 | 803 | 3500090 | 60 | |
| U49507.1 | - | 5 - 6 | 948 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lsr
Genomic Map