Assay Details
| Gene Symbol: Farsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025648.3 | NP_079924.2 | 3 - 4 | 428 | 82 | |
| GenBank mRNA | AK002577.1 | - | 3 - 4 | 405 | 82 | |
| AK031697.1 | - | 3 - 4 | 406 | 82 | ||
| AK044637.1 | - | 3 - 4 | 406 | 82 | ||
| AK084031.1 | - | 3 - 4 | 390 | 82 | ||
| AK154629.1 | - | 3 - 4 | 426 | 82 | ||
| AK171872.1 | - | 3 - 4 | 309 | 82 | ||
| BC006862.1 | - | 3 - 4 | 399 | 3595108 | 82 | |
| BC013533.1 | - | 3 - 4 | 397 | 4238911 | 82 | |
| CT010350.1 | - | 3 - 4 | 379 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Farsa
Genomic Map