Assay Details
| Gene Symbol: Farsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025648.3 | NP_079924.2 | 5 - 6 | 645 | 98 | |
| GenBank mRNA | AK002577.1 | - | 5 - 6 | 622 | 98 | |
| AK016506.1 | - | 5 - 6 | 641 | 98 | ||
| AK031697.1 | - | 5 - 6 | 623 | 98 | ||
| AK044637.1 | - | 5 - 6 | 623 | 98 | ||
| AK084031.1 | - | 5 - 6 | 607 | 98 | ||
| AK154629.1 | - | 5 - 6 | 643 | 98 | ||
| AK171872.1 | - | 5 - 6 | 525 | 97 | ||
| AK213018.1 | - | 1 - 2 | 109 | 98 | ||
| BC006862.1 | - | 5 - 6 | 616 | 3595108 | 98 | |
| BC013533.1 | - | 5 - 6 | 614 | 4238911 | 98 | |
| CT010350.1 | - | 5 - 6 | 596 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Farsa
Genomic Map