Assay Details
Gene Symbol: Nob1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026277.3 | NP_080553.1 | 1 - 2 | 73 | 68 | |
GenBank mRNA | AK021194.1 | - | 1 - 2 | 77 | 68 | |
AK054034.1 | - | 1 - 2 | 74 | 68 | ||
AK075755.1 | - | 1 - 2 | 74 | 68 | ||
AK165521.1 | - | 1 - 2 | 87 | 68 | ||
BC034900.1 | - | 1 - 2 | 74 | 68 | ||
BC103793.1 | - | 1 - 2 | 76 | 6328184 | 68 | |
BC117992.2 | - | 1 - 2 | 74 | 40104531 | 68 | |
BC117993.2 | - | 1 - 2 | 74 | 40104533 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
